Learn More
Waters Corp ACQUITY UPLC Protein SEC Column 450Å, 2.5 µm, 4.6 mm x 150 mm and BEH450 SEC Protein Standard Mix
Supplier:  Waters Corp 176002996
Encompass_Preferred
This column and standard kit includes the BEH450 SEC Protein Standard Mix (P/N 186006842) and an ACQUITY UPLC Protein SEC Column. Based upon the highly successful BEH particle technology, the ACQUITY UPLC BEH450 SEC 2.5 µm column is fully optimized for the analysis of proteins and their aggregates with molecular weights ranging from 100,000 to 1,500,000 Daltons, the total system solution will routinely deliver fast, accurate aggregate determinations up to 10 times quicker that traditional HPLC-based SEC methods..
Catalog No. 50-465-950