Learn More
Waters Corp Waters ACQUITY UPLC Method Development Kit, 2.1 × 100 mm, 1.7 μm, 4/pk
Supplier:  Waters Corp 176001043
Encompass_Preferred
This ACQUITY UPLC method development kit is designed for high and low pH applications, offering the widest selectivities for UPLC columns.
  • Column chemistries include BEH C18, BEH C8, BEH Shield RP18, and BEH Phenyl.
  • Particle size is 1.7 μm (BEH).
  • Dimensions are 2.1 × 100 mm.
  • Kit contains 4 columns per package.
  • Maximizes separation selectivity by exploring low and high mobile-phase pH conditions.
Catalog No. 50-780-018
Add to Cart
Request a Quote