Learn More
Waters Corp TUV Flow Cell, Titanium, 5 mm Pathlength, 1500 nL
Catalog No. NC1956838 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This TUV flow cell is designed for use with TUV detectors. It features a titanium construction, a 5 mm pathlength, and a 1500 nL illuminated volume. It is recommended for protein samples.
  • Titanium material provides durability.
  • 5 mm pathlength for optimal detection.
  • 1500 nL illuminated volume for precise measurements.
  • Recommended for protein samples.
  • For use with tunable UV detectors.
Catalog No. NC1956838 Supplier Waters Corp Supplier No. 205000611
Add to Cart
Add to Cart
Request a Quote
This TUV flow cell is designed for use with TUV detectors. It features a titanium construction, a 5 mm pathlength, and a 1500 nL illuminated volume. It is recommended for protein samples.
  • Titanium material provides durability.
  • 5 mm pathlength for optimal detection.
  • 1500 nL illuminated volume for precise measurements.
  • Recommended for protein samples.
  • For use with tunable UV detectors.