Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Waters ACQUITY UPLC Qualification Standards Kit, UV, ELSD, 0.1600 mg/mL Caffeine

Catalog No. 50201671 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

This Waters ACQUITY UPLC Qualification Standards Kit provides essential standards for UPLC systems, particularly those incorporating an Evaporative Light Scattering Detector (ELSD). Each kit includes a single 10 mL ampule of standard solution, formulated with 0.1600 mg/mL Caffeine with a precision of ±0.5%, designed for system performance applications.

  • Contains 1 ampule of standard solution.
  • Volume of 10 mL.
  • Concentration of 0.1600 mg/mL Caffeine ±0.5%.
  • Suitable for reversed phase separation mode.
  • Supports UV and ELSD detection methods.
  • Formulated as a liquid.
  • Shelf life of 36 months.
  • Shipped under ambient conditions.
  • Stored at room temperature.
  • Designed for system performance applications.

Catalog No. 50-201-671 Supplier Waters Corp Supplier No. 700002848
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This Waters ACQUITY UPLC Qualification Standards Kit provides essential standards for UPLC systems, particularly those incorporating an Evaporative Light Scattering Detector (ELSD). Each kit includes a single 10 mL ampule of standard solution, formulated with 0.1600 mg/mL Caffeine with a precision of ±0.5%, designed for system performance applications.

  • Contains 1 ampule of standard solution.
  • Volume of 10 mL.
  • Concentration of 0.1600 mg/mL Caffeine ±0.5%.
  • Suitable for reversed phase separation mode.
  • Supports UV and ELSD detection methods.
  • Formulated as a liquid.
  • Shelf life of 36 months.
  • Shipped under ambient conditions.
  • Stored at room temperature.
  • Designed for system performance applications.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.