Learn More
Waters Corp Waters Standards Kit, Reversed-Phase, UV Detection, 10 mL
Catalog No. 50201666 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The ACQUITY UPLC / UV Start-up Standards Kit includes two 10 mL ampules of reversed-phase standard solutions. Designed for UPLC/UV system performance verification and calibration, these standards aid in assessing reproducibility, flow rate, gradient performance, and detector/injector accuracy. They have a 24-month shelf life at room temperature.
  • Verifies system reproducibility.
  • Verifies flow rate and gradient performance.
  • Evaluates detector linearity and accuracy.
  • Assesses injector accuracy and carryover.
  • For system calibration.
  • Verifies system performance.
  • Reversed-phase standard.
  • Shelf life of 24 months.
  • Ships 1 per package.
  • Each solution contains 10 mL.
Catalog No. 50-201-666 Supplier Waters Corp Supplier No. 700002669
The ACQUITY UPLC / UV Start-up Standards Kit includes two 10 mL ampules of reversed-phase standard solutions. Designed for UPLC/UV system performance verification and calibration, these standards aid in assessing reproducibility, flow rate, gradient performance, and detector/injector accuracy. They have a 24-month shelf life at room temperature.
  • Verifies system reproducibility.
  • Verifies flow rate and gradient performance.
  • Evaluates detector linearity and accuracy.
  • Assesses injector accuracy and carryover.
  • For system calibration.
  • Verifies system performance.
  • Reversed-phase standard.
  • Shelf life of 24 months.
  • Ships 1 per package.
  • Each solution contains 10 mL.