Learn More
Waters Corp Waters Column, 300Å, 1.7 μm, 2.1 × 150 mm
Supplier:  Waters Corp 176003702
Encompass_Preferred
ACQUITY Glycoprotein BEH Amide 300Å, 1.7 μm columns are designed for analyzing intact glycoproteins, fragments, and glycopeptides. Each batch is quality control tested with Glycoprotein Performance Test Standard for batch-to-batch consistency. The kit includes one column and one vial of standard for immediate benchmarking upon receipt.
  • Packing material made with extremely pure reagents in a cGMP, ISO 9001 certified facility.
  • Each batch of column is qualified using standards, with results held to limited specification ranges for repeatable performance.
  • Every column is efficiency evaluated, and an eCord with a Certificate of Analysis and Performance Test Chromatogram is attached.
  • This information is also available to users of non-LC systems without an eCord reader.
Catalog No. 50-138-4496