Learn More
Waters Corp Waters Corporation SEC Column, 200 Å, 1.7 μm, 2.1 mm, 150 mm
Catalog No. 501384903 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This SEC column utilizes BEH particle technology, offering the first commercially available sub-2-μm SEC chemistry. Optimized for proteins and their aggregates (10,000 to 450,000 Daltons), it delivers fast, accurate aggregate determinations up to 10 times quicker than traditional HPLC methods. Designed for UPLC SEC applications, it ensures optimal separation performance with low LC system volume.
  • BEH particle technology.
  • Sub-2-μm SEC chemistry.
  • Analyzes proteins and aggregates (10,000 to 450,000 Daltons).
  • Up to 10x faster aggregate determination than HPLC.
  • Optimized for UPLC SEC applications.
  • Supports gel filtration and size exclusion chromatography.
  • Suitable for aggregate, monomer, and fragment analysis.
  • Compatible with various mobile phases.
  • Offers quantitative protein analysis with reproducible results.
This item is not returnable. View return policy
Catalog No. 50-138-4903 Supplier Waters Corp Supplier No. 186008471
Add to Cart
Add to Cart
This SEC column utilizes BEH particle technology, offering the first commercially available sub-2-μm SEC chemistry. Optimized for proteins and their aggregates (10,000 to 450,000 Daltons), it delivers fast, accurate aggregate determinations up to 10 times quicker than traditional HPLC methods. Designed for UPLC SEC applications, it ensures optimal separation performance with low LC system volume.
  • BEH particle technology.
  • Sub-2-μm SEC chemistry.
  • Analyzes proteins and aggregates (10,000 to 450,000 Daltons).
  • Up to 10x faster aggregate determination than HPLC.
  • Optimized for UPLC SEC applications.
  • Supports gel filtration and size exclusion chromatography.
  • Suitable for aggregate, monomer, and fragment analysis.
  • Compatible with various mobile phases.
  • Offers quantitative protein analysis with reproducible results.