Learn More
Waters Corp WATERS CORPORATION ACQUITY UPLC CSH Fluoro-Phenyl Method Validation Kit, 1.7 μm, 2.1 x 150 mm, 3/pk
Catalog No. 50201504 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The ACQUITY UPLC Method Validation Kit provides columns for evaluating the long-term reliability and robustness of analytical methods. This kit features CSH Fluoro-Phenyl chemistry, ensuring consistent performance for critical applications.
  • Includes three batches of chromatographic media from different base particles to evaluate method quality, reliability, and consistency.
  • Features CSH Fluoro-Phenyl column chemistry.
  • Equipped with a 1.7 μm particle size.
  • Dimensions of 2.1 × 150 mm.
  • Contains 3 columns per pack.
Catalog No. 50-201-504 Supplier Waters Corp Supplier No. 186006018
The ACQUITY UPLC Method Validation Kit provides columns for evaluating the long-term reliability and robustness of analytical methods. This kit features CSH Fluoro-Phenyl chemistry, ensuring consistent performance for critical applications.
  • Includes three batches of chromatographic media from different base particles to evaluate method quality, reliability, and consistency.
  • Features CSH Fluoro-Phenyl column chemistry.
  • Equipped with a 1.7 μm particle size.
  • Dimensions of 2.1 × 150 mm.
  • Contains 3 columns per pack.