Learn More
Waters Corp Waters Column, Amide, 1.7 μm, 300 Å, 2.1 mm X 150 mm
Supplier: Waters Corp 186007963
This column is designed for intact glycoprotein profiling, subunit fragment analysis, and glycopeptide mapping via hydrophilic interaction chromatography (HILIC). It delivers high-speed, high-resolution analysis for complex matrices, utilizing 1.7 μm particle size and BEH technology with an amide phase for extended column life.
- Optimized for glycoprotein profiling and analysis.
- Batch quality control tested for consistent performance.
- Manufactured in a cGMP, ISO 9001 certified facility.
- High-speed, high-resolution carbohydrate analysis.
- Prevents Schiff base formation, improving quantitation accuracy.
- High pH stability for enhanced MS detection and reduced run times.
- Operating pH range: 2–10.
- Temperature range: 20°C to 90°C.
- Pressures up to 15,000 psi tolerated.
- eCord intelligent chip for data tracking.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.