Learn More
Aviva Systems Biology Corp ACR Antibody - C-terminal region (ARP63016_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP63016_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ACR. It was validated on Western Blot. . Alternate names: -ApplicationWB. Species reactivity: Cow, Horse, Human, Sheep
Catalog No. 50-287-846