Learn More
Cayman Chemical AcrBEcolI K12 10ug
SDP
Catalog No. 503015841
Encompass_Preferred
Source: C-terminally His-tagged AcrB (E. coli K12 recombinant) purified from E. coli • AAs: full length, wild-type sequence • MW: 114.6 kDa
Catalog No. 50-301-5841 Supplier Cayman Chemical Supplier No. 2473310 UG
Source: C-terminally His-tagged AcrB (E. coli K12 recombinant) purified from E. coli • AAs: full length, wild-type sequence • MW: 114.6 kDa