Learn More
Cytiva Pall Acrodisc Syringe Filters with PVDF Membrane - 0.2 µm, 25mm (50/pkg)
Supplier:  Cytiva 4406TC
Encompass
Acrodisc Syringe Filters with PVDF Membrane - 0.2 µm, Standard 25mm.Syringe filters with PVDF membrane are ideal for the filtration of many organic and non-organic solvents in HPLC (0.45 μm) and UHPLC (0.2 μm) sample preparation. Filtration helps protect sensitive parts of analytical instrumentation from particulate build-up, extending chromatography column life and reducing system downtime.PVDF syringe filters are chemical compatibility with aggressive acids and alcohols. They are not recommended for the filtration of acetone, DMF, DMSO, or strong bases.25 mm Standard Acrodisc Syringe Filters are ideal for sample volumes of ≤ 100 mL and feature an effective filtration area of 2.8 cm2. They feature a polypropylene housing that is chemically resistant to harsh solvents and low in extractables. This product is not approved for use in medical, clinical, surgical or other patient applications.
Catalog No. NC1963926