Learn More
Cayman Chemical AcremonIum AselacIn C 100ug
SDP
Catalog No. 502950444
Encompass_Preferred
A cyclic pentapeptolide antagonist of ETA and ETB; inhibits the binding of endothelin-1 to isolated bovine atrial membranes and porcine cerebral membranes (IC50s = 60 and 80 UG/ml, respectively)
Catalog No. 50-295-0444 Supplier Cayman Chemical Supplier No. 33163100 UG
A cyclic pentapeptolide antagonist of ETA and ETB; inhibits the binding of endothelin-1 to isolated bovine atrial membranes and porcine cerebral membranes (IC50s = 60 and 80 UG/ml, respectively)