Learn More
Medchemexpress LLC Acridin-9-amine hydrochloride hydrate | 52417-22-8 | 248.71 | 100 G
SDP
Catalog No. 5000452852
Encompass_Preferred
Acridin-9-amine (9-Aminoacridine) hydrochloride hydrate is a mutagen and DNA modifier.
  • Induces mutagenic lesions during DNA replication.
Catalog No. 50-004-52852 Supplier Medchemexpress LLC Supplier No. HYW011950100G
Acridin-9-amine (9-Aminoacridine) hydrochloride hydrate is a mutagen and DNA modifier.
  • Induces mutagenic lesions during DNA replication.