Learn More
Medchemexpress LLC Acridin-9-amine hydrochloride hydrate | 52417-22-8 | 248.71 | 50 G
SDP
Catalog No. 5000452760
Encompass_Preferred
Acridin-9-amine (9-Aminoacridine) hydrochloride hydrate is a mutagen and DNA modifier that induces mutagenic lesions during DNA replication.
Catalog No. 50-004-52760 Supplier Medchemexpress LLC Supplier No. HYW01195050G
Acridin-9-amine (9-Aminoacridine) hydrochloride hydrate is a mutagen and DNA modifier that induces mutagenic lesions during DNA replication.