Learn More
Medchemexpress LLC Acridin-9-amine hydrochloride hydrate | 52417-22-8 | >98% | 248.71 | 1 G
SDP
Catalog No. 5000452230
Encompass_Preferred
Acridin-9-amine (9-Aminoacridine) hydrochloride hydrate is a mutagen and DNA modifier. It induces mutagenic lesions during DNA replication.
  • Mutagen
  • DNA modifier
  • Induces mutagenic lesions during DNA replication
Catalog No. 50-004-52230 Supplier Medchemexpress LLC Supplier No. HYW0119501G
Acridin-9-amine (9-Aminoacridine) hydrochloride hydrate is a mutagen and DNA modifier. It induces mutagenic lesions during DNA replication.
  • Mutagen
  • DNA modifier
  • Induces mutagenic lesions during DNA replication