Learn More
Medchemexpress LLC Acridin-9-amine hydrochloride hydrate | 52417-22-8 | MFCD00150071 | 248.71 | 25 G
SDP
Catalog No. 5000452675
Encompass_Preferred
Acridin-9-amine (9-Aminoacridine) hydrochloride hydrate is a mutagen and DNA modifier that induces mutagenic lesions during DNA replication. It is intended for research use only.
  • Mutagen
  • DNA modifier
  • Induces mutagenic lesions during DNA replication
  • For research use only
Catalog No. 50-004-52675 Supplier Medchemexpress LLC Supplier No. 25GHYW011950
Acridin-9-amine (9-Aminoacridine) hydrochloride hydrate is a mutagen and DNA modifier that induces mutagenic lesions during DNA replication. It is intended for research use only.
  • Mutagen
  • DNA modifier
  • Induces mutagenic lesions during DNA replication
  • For research use only