Learn More
Medchemexpress LLC Acridin-9-amine hydrochloride hydrate | 52417-22-8 | MFCD00150071 | 98% | 248.71 | 5 G
SDP
Catalog No. 5000453480
Encompass_Preferred
Acridin-9-amine (9-Aminoacridine) hydrochloride hydrate is a mutagen and DNA modifier. It induces mutagenic lesions during DNA replication. This product is intended for research use only and is not sold to patients.
  • Mutagen
  • DNA modifier
  • Induces mutagenic lesions during DNA replication
  • For research use only
Catalog No. 50-004-53480 Supplier Medchemexpress LLC Supplier No. HYW0119505G
Acridin-9-amine (9-Aminoacridine) hydrochloride hydrate is a mutagen and DNA modifier. It induces mutagenic lesions during DNA replication. This product is intended for research use only and is not sold to patients.
  • Mutagen
  • DNA modifier
  • Induces mutagenic lesions during DNA replication
  • For research use only