Learn More
Cayman Chemical Acridine Orange 10-nonyl (bromide), Purity: ≥90%, CAS: 75168-11-5, 5 mg
SDP
Catalog No. 503015844
Encompass_Preferred
A mitochondrial dye; incorporates into the cardiolipin-rich inner mitochondrial membrane; selectively binds to cardiolipin-containing liposomes over phosphatidylserine- and phosphatidylinositol-containing liposomes in vitro (Kas = 0.002, 7,000, and 7,000 mM, respectively); binding dependent on mitochondrial membrane potential; ex/em = 490/510 nm, respectively; has been used to quantify cardiolipin content and mitochondrial mass in isolated mitochondria and cellular systems
Catalog No. 50-301-5844 Supplier Cayman Chemical Supplier No. 255455 MG
A mitochondrial dye; incorporates into the cardiolipin-rich inner mitochondrial membrane; selectively binds to cardiolipin-containing liposomes over phosphatidylserine- and phosphatidylinositol-containing liposomes in vitro (Kas = 0.002, 7,000, and 7,000 mM, respectively); binding dependent on mitochondrial membrane potential; ex/em = 490/510 nm, respectively; has been used to quantify cardiolipin content and mitochondrial mass in isolated mitochondria and cellular systems