Learn More
Cayman Chemical AcrIdIn Ornge 10nonylbro 100mg
SDP
Catalog No. 503015914
Encompass_Preferred
A mitochondrial dye; incorporates into the cardiolipin-rich inner mitochondrial membrane; selectively binds to cardiolipin-containing liposomes over phosphatidylserine- and phosphatidylinositol-containing liposomes in vitro (Kas = 0.002, 7,000, and 7,000 mM, respectively); binding dependent on mitochondrial membrane potential; ex/em = 490/510 nm, respectively; has been used to quantify cardiolipin content and mitochondrial mass in isolated mitochondria and cellular systems
Catalog No. 50-301-5914 Supplier Cayman Chemical Supplier No. 25545100 MG
A mitochondrial dye; incorporates into the cardiolipin-rich inner mitochondrial membrane; selectively binds to cardiolipin-containing liposomes over phosphatidylserine- and phosphatidylinositol-containing liposomes in vitro (Kas = 0.002, 7,000, and 7,000 mM, respectively); binding dependent on mitochondrial membrane potential; ex/em = 490/510 nm, respectively; has been used to quantify cardiolipin content and mitochondrial mass in isolated mitochondria and cellular systems