Learn More
Cayman Chemical Acridine Orange, CAS: 494-38-2, 5 g
SDP
Catalog No. 502952666
Encompass_Preferred
A fluorescent cationic dye; ex/em spectra = 502/525, 460/650, and 475/590 nm when bound to dsDNA, bound to ssDNA or RNA, and under acidic conditions, respectively; ratio of emission at 590 to emission at 525 nm can be used to quantify the increase in acidic vesicular organelles observed during cellular autophagy
Catalog No. 50-295-2666 Supplier Cayman Chemical Supplier No. 143385 G
A fluorescent cationic dye; ex/em spectra = 502/525, 460/650, and 475/590 nm when bound to dsDNA, bound to ssDNA or RNA, and under acidic conditions, respectively; ratio of emission at 590 to emission at 525 nm can be used to quantify the increase in acidic vesicular organelles observed during cellular autophagy