Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acridine Hydrochloride Hydrate 98.0+%, TCI America™
SDP

Catalog No. A01315G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A01315G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A01315G Supplier TCI America Supplier No. A01315G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 17784-47-3
Molecular Formula C13H10ClN
Molecular Weight (g/mol) 215.68
MDL Number MFCD00035149
InChI Key XUESTGHCVFYOLL-UHFFFAOYSA-N
Synonym acridine hydrochloride, acridine monohydrochloride, acmc-1c7yu, acridinehydrochloridehydrate, c13h9n.hcl, cambridge id 5510890, acridine, hydrochloride 1:1
PubChem CID 2849400
IUPAC Name acridine;hydrochloride
SMILES C1=CC=C2C(=C1)C=C3C=CC=CC3=N2.Cl

Specifications

Quantity 5 g
Formula Weight 215.68
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Acridine Hydrochloride Hydrate
RTECSNumber AR9322000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.