Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acridine hydrochloride, Thermo Scientific™

Catalog No. AAJ6263403
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6263403 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ6263403 Supplier Thermo Scientific Chemicals Supplier No. J6263403

missing translation for 'validMainframeMsg'

Acid fast bacillus stain

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Acridine hydrochloride is a fluorescent compound used to investigate the release properties of multilayer nanocapsules. Acridine hydrochloride has been utilized as a hydrophilic drug model and was loaded into the nano-structures.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17784-47-3
Molecular Formula C13H10ClN
Molecular Weight (g/mol) 215.68
MDL Number MFCD00035149
InChI Key XUESTGHCVFYOLL-UHFFFAOYSA-N
Synonym acridine hydrochloride, acridine monohydrochloride, acmc-1c7yu, acridinehydrochloridehydrate, c13h9n.hcl, cambridge id 5510890, acridine, hydrochloride 1:1
PubChem CID 2849400
IUPAC Name acridine;hydrochloride
SMILES C1=CC=C2C(=C1)C=C3C=CC=CC3=N2.Cl

Specifications

Color Yellow to Orange
Quantity 1 g
Beilstein 3697519
Formula Weight 215.68
Physical Form Powder
Chemical Name or Material Acridine hydrochloride
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
RTECSNumber AR9322000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.