Learn More
Medchemexpress LLC HY-D0993 10mg Medchemexpress, Acridine Orange 10-Nonyl Bromide CAS:75168-11-5 Purity:>98%
SDP
Catalog No. 501933230
Encompass_Preferred
Medchemexpress, HY-D0993 10mg Acridine Orange 10-Nonyl Bromide CAS:75168-11-5 Acridine Orange 10-Nonyl Bromide is a fluorescent probe for cardiolipin (λ ex : 489 nm, λ em : 525 nm). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3230 Supplier Medchemexpress LLC Supplier No. HYD099310MG
Medchemexpress, HY-D0993 10mg Acridine Orange 10-Nonyl Bromide CAS:75168-11-5 Acridine Orange 10-Nonyl Bromide is a fluorescent probe for cardiolipin (λ ex : 489 nm, λ em : 525 nm). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.