Learn More
AdipoGen Acridine orange 10-nonyl Br
SDP
Catalog No. 502561834
Encompass_Preferred
Chemical. CAS 75168-11-5. Formula C26H38BrN3. MW 472.5. Acridine orange 10-nonyl is a green fluorescent mitochondrial dye. The dye incorporates into the cardiolipin-rich inner mitochondrial membrane. Acridine orange 10-nonyl has been used to study multidrug resistance and to measure changes in mitochondrial mass during apoptosis. Acridine orange 10-nonyl has been used to quantify cardiolipin content and mitochondrial mass in isolated mitochondria and cellular systems. Spectral Data lambdaEx/lambdaEm MeOH = 495/522nm.
Catalog No. 50-256-1834 Supplier AdipoGen Supplier No. CDXA0103M100
Chemical. CAS 75168-11-5. Formula C26H38BrN3. MW 472.5. Acridine orange 10-nonyl is a green fluorescent mitochondrial dye. The dye incorporates into the cardiolipin-rich inner mitochondrial membrane. Acridine orange 10-nonyl has been used to study multidrug resistance and to measure changes in mitochondrial mass during apoptosis. Acridine orange 10-nonyl has been used to quantify cardiolipin content and mitochondrial mass in isolated mitochondria and cellular systems. Spectral Data lambdaEx/lambdaEm MeOH = 495/522nm.