Learn More
Medchemexpress LLC Acridine orange 10-nonyl bromide | 75168-11-5 | 99.5% | 472.50 | 25 MG
SDP
Catalog No. 5000429519
Encompass_Preferred
Acridine Orange 10-Nonyl Bromide is a fluorescent probe for cardiolipin with an excitation wavelength (λex) of 489 nm and an emission wavelength (λem) of 525 nm. It is used for research purposes only.
  • Fluorescent probe for cardiolipin.
  • Can be used to quantify cardiolipin in isolated mitochondria.
  • At 4.75 μM, it saturates mitochondrial binding sites in 1.5 X 10^6 cells within 15 minutes at 20°C.
  • Upon interaction with cardiolipin, the dye's excitation and emission wavelengths shift from 496 nm and 525 nm to 450 nm and 640 nm, respectively.
  • Increasing amounts of cardiolipin (0 to 30 μM) and other acidic phospholipids in thin-walled vesicles, when added to 45 μM Acridine Orange 10-Nonyl Bromide, changes the red fluorescence emission measured at 640 nm according to the liposome composition.
Catalog No. 50-004-29519 Supplier Medchemexpress LLC Supplier No. 25MGHYD0993
Acridine Orange 10-Nonyl Bromide is a fluorescent probe for cardiolipin with an excitation wavelength (λex) of 489 nm and an emission wavelength (λem) of 525 nm. It is used for research purposes only.
  • Fluorescent probe for cardiolipin.
  • Can be used to quantify cardiolipin in isolated mitochondria.
  • At 4.75 μM, it saturates mitochondrial binding sites in 1.5 X 10^6 cells within 15 minutes at 20°C.
  • Upon interaction with cardiolipin, the dye's excitation and emission wavelengths shift from 496 nm and 525 nm to 450 nm and 640 nm, respectively.
  • Increasing amounts of cardiolipin (0 to 30 μM) and other acidic phospholipids in thin-walled vesicles, when added to 45 μM Acridine Orange 10-Nonyl Bromide, changes the red fluorescence emission measured at 640 nm according to the liposome composition.