Learn More
Electron Microscopy Sciences Acridine Orange 25 GR
SDP
Catalog No. 50980158
Encompass_Preferred
(Basic Orange 14)
CAS #10127-02-3 C₁₇H₂ON₃Cl F.W. 301.82
Ultrastructural probes on DNA templates within human bone marrow and lymph node cells (Frenstar, J., Adv. Cell Mol. Biol., 3, 11974). Used to detect changes in DNA template activity within cells (Frenster, J. Cell Nucl., 1, 565 1974). And used as a fluorescent dye for cytochemistry (J. Histochem. Cytochem., 31, 737 1983).
Catalog No. 50-980-158 Supplier Electron Microscopy Sciences Supplier No. 10050
(Basic Orange 14)
CAS #10127-02-3 C₁₇H₂ON₃Cl F.W. 301.82
Ultrastructural probes on DNA templates within human bone marrow and lymph node cells (Frenstar, J., Adv. Cell Mol. Biol., 3, 11974). Used to detect changes in DNA template activity within cells (Frenster, J. Cell Nucl., 1, 565 1974). And used as a fluorescent dye for cytochemistry (J. Histochem. Cytochem., 31, 737 1983).