Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Acridine Orange, pure, ca. 55% dye content

Catalog No. ac300911000
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC300911000 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC300911000 Supplier Thermo Scientific Chemicals Supplier No. 300911000
Only null left
Add to Cart

CAS: 10127-02-3 | C17H20Cl3N3Zn | 438.10 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10127-02-3
Molecular Formula C17H20Cl3N3Zn
Molecular Weight (g/mol) 438.10
MDL Number MFCD00081043,MFCD00081043,MFCD00081043
InChI Key VADJQOXWNSPOQA-UHFFFAOYSA-L
Synonym Basic Orange 14, 3,6-Bis(dimethylamino)acridine zinc chloride double salt, C.I. 46005
PubChem CID 71774334
IUPAC Name zinc;hydron;3-N,3-N,6-N,6-N-tetramethylacridine-3,6-diamine;trichloride
SMILES [H+].[Cl-].[Cl-].[Cl-].[Zn++].CN(C)C1=CC2=NC3=CC(=CC=C3C=C2C=C1)N(C)C

Specifications

Melting Point 165.0°C
Dye Content 50% min.
Solubility Information Solubility in water: soluble.
Infrared Spectrum Authentic
Formula Weight 369.94
Grade Pure
Quantity 100 g
Chemical Name or Material Acridine Orange
Hazard Category Warning
Hazard Statement GHS H Statement Harmful if swallowed. Harmful if inhaled. Harmful in contact with skin. Suspected of causing genetic defects.
Precautionary Statement GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. Use personal protective equipment as required. IF ON SKIN: Wash with plenty of soap and water. Do not breathe dust/fume/gas/mist/vapors/spray. IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.