Learn More
Medchemexpress LLC Acridine orange base | 494-38-2 | 96.58% | 100 MG
SDP
Catalog No. 5000409951
Encompass_Preferred
Acridine Orange base is a cell-permeable fluorescent dye utilized for staining various organisms such as bacteria, parasites, and viruses a bright orange. When applied under specific conditions (pH=3.5, Ex=460 nm), it allows for the distinction of human cells, which appear green, through fluorescence microscopy. This dye exhibits green fluorescence when bound to double-stranded DNA (dsDNA) and red fluorescence when bound to single-stranded DNA (ssDNA) or single-stranded RNA (ssRNA), making it suitable for cell cycle assays.
  • Cell-permeable fluorescent dye.
  • Stains organisms bright orange.
  • Distinguishes human cells in green.
  • Fluoresces green with dsDNA.
  • Fluoresces red with ssDNA or ssRNA.
  • Useful for cell cycle assays.
Catalog No. 50-004-09951 Supplier Medchemexpress LLC Supplier No. HYD0952100MG
Acridine Orange base is a cell-permeable fluorescent dye utilized for staining various organisms such as bacteria, parasites, and viruses a bright orange. When applied under specific conditions (pH=3.5, Ex=460 nm), it allows for the distinction of human cells, which appear green, through fluorescence microscopy. This dye exhibits green fluorescence when bound to double-stranded DNA (dsDNA) and red fluorescence when bound to single-stranded DNA (ssDNA) or single-stranded RNA (ssRNA), making it suitable for cell cycle assays.
  • Cell-permeable fluorescent dye.
  • Stains organisms bright orange.
  • Distinguishes human cells in green.
  • Fluoresces green with dsDNA.
  • Fluoresces red with ssDNA or ssRNA.
  • Useful for cell cycle assays.