Learn More
Medchemexpress LLC Acridine Orange base | 494-38-2 | 96.6% | 250 MG
SDP
Catalog No. 5000414118
Encompass_Preferred
Acridine Orange base is a cell-permeable fluorescent dye that stains organisms (bacteria, parasites, viruses, etc.) bright orange. When used under appropriate conditions (pH=3.5, Ex=460 nm), Acridine Orange base distinguishes human cells in green for detection by fluorescence microscopy. It fluoresces green when bound to dsDNA (Ex=488, Em=520-524) and red when bound to ssDNA (Ex=457, Em=630-644) or ssRNA (Ex=457, Em=630-644). It can also be used in cell cycle assays.
  • Cell-permeable fluorescent dye
  • Stains various organisms bright orange
  • Distinguishes human cells in green under specific conditions
  • Fluoresces green when bound to dsDNA
  • Fluoresces red when bound to ssDNA or ssRNA
  • Can be used in cell cycle assays
Catalog No. 50-004-14118 Supplier Medchemexpress LLC Supplier No. 250MGHYD0952
Acridine Orange base is a cell-permeable fluorescent dye that stains organisms (bacteria, parasites, viruses, etc.) bright orange. When used under appropriate conditions (pH=3.5, Ex=460 nm), Acridine Orange base distinguishes human cells in green for detection by fluorescence microscopy. It fluoresces green when bound to dsDNA (Ex=488, Em=520-524) and red when bound to ssDNA (Ex=457, Em=630-644) or ssRNA (Ex=457, Em=630-644). It can also be used in cell cycle assays.
  • Cell-permeable fluorescent dye
  • Stains various organisms bright orange
  • Distinguishes human cells in green under specific conditions
  • Fluoresces green when bound to dsDNA
  • Fluoresces red when bound to ssDNA or ssRNA
  • Can be used in cell cycle assays