Learn More
Medchemexpress LLC Acridine orange (euchrysine 3RX) zinc chloride salt | 10127-02-3 | MFCD00081043 | 90% | 100 MG
SDP
Catalog No. 5000410211
Encompass_Preferred
Acridine Orange (Euchrysine 3RX) zinc chloride salt is a cell-penetrable nucleic acid-selective fluorescent dye. It produces orange fluorescence when binding to single-stranded DNA (ssDNA) or RNA, and green fluorescence when binding to double-stranded DNA (dsDNA). The excitation is at 488 nM, with green fluorescence emitted at 530 nm and orange fluorescence at 640 nm.
  • Cell-penetrable nucleic acid-selective fluorescent dye.
  • Produces orange fluorescence with ssDNA or RNA.
  • Produces green fluorescence with dsDNA.
  • Excitation at 488 nM.
  • Green fluorescence emission at 530 nm.
  • Orange fluorescence emission at 640 nm.
Catalog No. 50-004-10211 Supplier Medchemexpress LLC Supplier No. HYD0942100MG
Acridine Orange (Euchrysine 3RX) zinc chloride salt is a cell-penetrable nucleic acid-selective fluorescent dye. It produces orange fluorescence when binding to single-stranded DNA (ssDNA) or RNA, and green fluorescence when binding to double-stranded DNA (dsDNA). The excitation is at 488 nM, with green fluorescence emitted at 530 nm and orange fluorescence at 640 nm.
  • Cell-penetrable nucleic acid-selective fluorescent dye.
  • Produces orange fluorescence with ssDNA or RNA.
  • Produces green fluorescence with dsDNA.
  • Excitation at 488 nM.
  • Green fluorescence emission at 530 nm.
  • Orange fluorescence emission at 640 nm.