Learn More
Abcam Acridine orange hemi(zinc chloride) salt, Fluorescent cationic DNA and RNA dye, 10G
Catalog No. 502704695
Encompass_Preferred
MW 369.96 g/mol. Cell-permeable metachromatic fluorescent cationic dye that stains DNA and RNA. Also an inhibitor of RNA polymerase.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-4695 Supplier Abcam Supplier No. AB14634810G
MW 369.96 g/mol. Cell-permeable metachromatic fluorescent cationic dye that stains DNA and RNA. Also an inhibitor of RNA polymerase.
The product is subject to the following: Abcam Restricted Use Statement