Learn More
Novus Biologicals™ Acridine Orange Hydrochloride

Description
Biological Activity: Cell and organelle membrane permeable nucleic acid binding dye. Emits green fluorescence when bound to double stranded DNA and red fluorescence when bound to RNA or single stranded DNA. Used in cell cycle and apoptosis studies and as a lysosomal dye.
Storage: Store at 4°C.
Note: Inhibitors are guranteed for 1 year from date of receipt.
Specifications
Specifications
| Description | CAS: 65-61-2 |
| Formulation | C17H19N3.HCl |
| For Use With (Application) | Inhibits apoptosis |
| Quantity | 50 mg |
| Product Type | Inhibitors |
| Molecular Weight (g/mol) | 301.81g/mol |
| Solubility | Soluble to 100mM in water, 100mM in DMSO |
| Content And Storage | -20°C |
| Target | Acridine Orange |
| Color | Dark Orange |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.