Learn More
Apexbio Technology LLC Acridine Orange hydrochloride 65-61-2 50mg
SDP
Supplier:  Apexbio Technology LLC B774750
Encompass_Preferred
Acridine Orange hydrochloride (65-61-2) is a small-molecule fluorochrome targeting nucleic acids It is designed to bind double-stranded DNA and single-stranded DNA or RNA thereby enabling differential nucleic acid staining in live cells Acridine Orange hydrochloride exerts its biological activity primarily through selective intercalation with nucleic acids and pH-dependent accumulation in acidic organelles resulting in green fluorescence for dsDNA and red/orange fluorescence for ssDNA/RNA In cell-based assays Acridine Orange hydrochloride exhibits cytotoxicity with reported IC50 values typically ranging from 0 5 5 M Based on these pharmacological properties Acridine Orange hydrochloride holds research potential in apoptosis detection assessment of lysosomal integrity and function and antitumor activity studies
Catalog No. 50-000-91193