Learn More
Medchemexpress LLC Acridine orange hydrochloride | 65-61-2 | MFCD00150070 | 99.7% | 301.81 g/mol | C17H20ClN3 | 200 MG
SDP
Catalog No. 5000248034
Encompass_Preferred
Acridine Orange hydrochloride is a cell-permeable, nucleic-acid-selective fluorescent dye used to differentially stain double-stranded DNA, single-stranded DNA, and RNA. It produces distinct emission spectra depending on nucleic acid binding, enabling visualization and quantification of nucleic acid content in cells for microscopy and flow cytometry applications.
  • Cell-permeable, nucleic-acid-selective fluorescent dye.
  • Differential staining: green emission with dsDNA, orange/red with ssDNA and RNA.
  • Excitation around 488 nm; emissions near 530 nm (dsDNA) and 640 nm (ssDNA/RNA).
  • Suitable for fluorescence microscopy and flow cytometry workflows.
  • Available in solid and solution formats for flexible experimental use.
Catalog No. 50-002-48034 Supplier Medchemexpress LLC Supplier No. HY101879200MG
Acridine Orange hydrochloride is a cell-permeable, nucleic-acid-selective fluorescent dye used to differentially stain double-stranded DNA, single-stranded DNA, and RNA. It produces distinct emission spectra depending on nucleic acid binding, enabling visualization and quantification of nucleic acid content in cells for microscopy and flow cytometry applications.
  • Cell-permeable, nucleic-acid-selective fluorescent dye.
  • Differential staining: green emission with dsDNA, orange/red with ssDNA and RNA.
  • Excitation around 488 nm; emissions near 530 nm (dsDNA) and 640 nm (ssDNA/RNA).
  • Suitable for fluorescence microscopy and flow cytometry workflows.
  • Available in solid and solution formats for flexible experimental use.