Learn More
Medchemexpress LLC Acridine orange hydrochloride | 65-61-2 | MFCD00150070 | 99.7% | 301.81 g/mol | C17H20ClN3 | 500 MG
SDP
Catalog No. 5000245564
Encompass_Preferred
Acridine orange hydrochloride is a cell-permeable, nucleic-acid-selective fluorescent dye supplied as a hydrochloride salt powder. It intercalates with nucleic acids and emits different fluorescence depending on binding to DNA or RNA, making it useful for microscopy, flow cytometry, and nucleic acid detection. High-purity material is typically provided with batch documentation.
  • Cell-permeable nucleic acid stain for DNA and RNA.
  • Distinctive fluorescence: green for DNA, orange for RNA.
  • Suitable for fluorescence microscopy and flow cytometry.
  • High purity with batch-specific documentation.
  • Supplied as a stable hydrochloride salt powder, convenient for solution preparation.
Catalog No. 50-002-45564 Supplier Medchemexpress LLC Supplier No. HY101879500MG
Acridine orange hydrochloride is a cell-permeable, nucleic-acid-selective fluorescent dye supplied as a hydrochloride salt powder. It intercalates with nucleic acids and emits different fluorescence depending on binding to DNA or RNA, making it useful for microscopy, flow cytometry, and nucleic acid detection. High-purity material is typically provided with batch documentation.
  • Cell-permeable nucleic acid stain for DNA and RNA.
  • Distinctive fluorescence: green for DNA, orange for RNA.
  • Suitable for fluorescence microscopy and flow cytometry.
  • High purity with batch-specific documentation.
  • Supplied as a stable hydrochloride salt powder, convenient for solution preparation.