Learn More
Description
Acridine Orange is a metachromatic dye which can stain DNA, RNA and acid glycosaminoglycans. At low concentration it intercalates into DNA and precipitates RNA. However, at high concentration it denatures and precipitates both RNA and DNA. Acridine Orange is also used to analyze autophagy. It goes into acidic organelles in a pH-dependent manner. At neutral pH, acridine orange gives a green fluorescence and in acidic conditions, it accumulates in the acidic organelle giving a bright red fluorescence.
Specifications
Specifications
| Chemical Name or Material | Acridine Orange Hydrochloride |
| CAS | 65-61-2 |
| Concentration | 10 mg/mL |
| Color | Orange to Red |
| Quantity | 10 mL |
| Molecular Weight (g/mol) | 301.81 g/mol |
| Percent Purity | ≥95% |
| Grade | Histological |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.