Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acridine Orange, MP Biomedicals

Catalog No. ICN203423 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN203423 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN203423 Supplier MP Biomedicals, Inc Supplier No. 0520342305

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 65-61-2
Molecular Formula C17H20ClN3
Molecular Weight (g/mol) 301.82
MDL Number MFCD00012660
InChI Key ASKSWIKIIFUEOI-UHFFFAOYSA-N
PubChem CID 517204
ChEBI CHEBI:51739
IUPAC Name hydrogen N1,N1,N7,N7-tetramethylacridine-1,7-diamine chloride
SMILES [H+].[Cl-].CN(C)C1=CC=C2N=C3C=CC=C(N(C)C)C3=CC2=C1

Specifications

Formula Weight 301.82
Quantity 5 g
Chemical Name or Material Acridine Orange
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.