Learn More
Abcam Acridine Orange Staining Solution, 500UL
Catalog No. 502656118
Encompass_Preferred
Acridine Orange (AO) is a slightly cationic, lipophilic, fluorochrome stain capable of permeating cell and organelle membrane structure.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-265-6118 Supplier Abcam Supplier No. AB270791500UL
Acridine Orange (AO) is a slightly cationic, lipophilic, fluorochrome stain capable of permeating cell and organelle membrane structure.
The product is subject to the following: Abcam Restricted Use Statement