Learn More
Medchemexpress LLC Acridinium C2 NHS ester | 177332-37-5 | 92.1% | 632.56 | C29H23F3N2O9S | 1mg
SDP
Catalog No. 5000268754
Encompass_Preferred
Acridinium C2 NHS ester is a chemiluminescent label that is active for the development of protein and nucleic acid probes Acridinium C2 NHS ester can be used in bioanalysis and diagnostics to provide highly sensitive detection solutions Acridinium C2 NHS ester is widely used in medical research especially in the monitoring and analysis of biomolecules
Catalog No. 50-002-68754 Supplier Medchemexpress LLC Supplier No. HYW7770121MG
Acridinium C2 NHS ester is a chemiluminescent label that is active for the development of protein and nucleic acid probes Acridinium C2 NHS ester can be used in bioanalysis and diagnostics to provide highly sensitive detection solutions Acridinium C2 NHS ester is widely used in medical research especially in the monitoring and analysis of biomolecules