Learn More
Enzo Life Sciences Acridinium C2, NHS Ester (1mg). CAS: 177332-37-5
SDP
Catalog No. 502007082
Encompass_Preferred
Formula: C29H23F3N2O9S. MW: 632.56. Formulation: Solid. Solubility: Soluble in DMSO or methanol. Long Term Storage: -20°C.
Catalog No. 50-200-7082 Supplier Enzo Life Sciences Supplier No. ADI9060001
Formula: C29H23F3N2O9S. MW: 632.56. Formulation: Solid. Solubility: Soluble in DMSO or methanol. Long Term Storage: -20°C.