Learn More
Cayman Chemical Acridinium NHS ester, Purity: ≥95%, CAS: 177332-37-5, 50 mg
SDP
Catalog No. 502983103
Encompass_Preferred
The acridinium NHS ester can be used to label proteins and nucleic acids. The covalently bound acridinium NHS ester will produce chemiluminescence in the presence of hydrogen peroxide. Acridinium-labeled proteins can be used as a detection method in immunoassays.{2437,2438}
Catalog No. 50-298-3103 Supplier Cayman Chemical Supplier No. 20020050 MG
The acridinium NHS ester can be used to label proteins and nucleic acids. The covalently bound acridinium NHS ester will produce chemiluminescence in the presence of hydrogen peroxide. Acridinium-labeled proteins can be used as a detection method in immunoassays.{2437,2438}