Learn More
Cayman Chemical Acridinium NHS ester, Purity: ≥95%, CAS: 177332-37-5, 5 mg
SDP
Catalog No. 502984871
Encompass_Preferred
The acridinium NHS ester can be used to label proteins and nucleic acids. The covalently bound acridinium NHS ester will produce chemiluminescence in the presence of hydrogen peroxide. Acridinium-labeled proteins can be used as a detection method in immunoassays.{2437,2438}
Catalog No. 50-298-4871 Supplier Cayman Chemical Supplier No. 2002005 MG
The acridinium NHS ester can be used to label proteins and nucleic acids. The covalently bound acridinium NHS ester will produce chemiluminescence in the presence of hydrogen peroxide. Acridinium-labeled proteins can be used as a detection method in immunoassays.{2437,2438}