Learn More
Medchemexpress LLC Acridone | 578-95-0 | 195.22 | 25 G
SDP
Catalog No. 5000449984
Encompass_Preferred
Acridone is an organic compound based on the acridine skeleton, recognized for its diverse biological activities. This chemical exhibits antibacterial, antimalarial, antiviral, and antineoplastic properties. It has demonstrated efficacy in inhibiting the proliferation of cancer cells and modulating the expression of ATP binding cassette subfamily G member 2 (ABCG2).
  • Organic compound based on the acridine skeleton.
  • Exhibits antibacterial activity.
  • Exhibits antimalarial activity.
  • Exhibits antiviral activity.
  • Exhibits antineoplastic activity.
  • Inhibits proliferation of MDA-MB-231 cancer cells.
  • Blocks mRNA and protein expression of ATP binding cassette subfamily G member 2 (ABCG2).
Catalog No. 50-004-49984 Supplier Medchemexpress LLC Supplier No. 25GHYW007771
Acridone is an organic compound based on the acridine skeleton, recognized for its diverse biological activities. This chemical exhibits antibacterial, antimalarial, antiviral, and antineoplastic properties. It has demonstrated efficacy in inhibiting the proliferation of cancer cells and modulating the expression of ATP binding cassette subfamily G member 2 (ABCG2).
  • Organic compound based on the acridine skeleton.
  • Exhibits antibacterial activity.
  • Exhibits antimalarial activity.
  • Exhibits antiviral activity.
  • Exhibits antineoplastic activity.
  • Inhibits proliferation of MDA-MB-231 cancer cells.
  • Blocks mRNA and protein expression of ATP binding cassette subfamily G member 2 (ABCG2).