Learn More
Medchemexpress LLC Acridone | 578-95-0 | 195.22 | 500 G
SDP
Supplier:  Medchemexpress LLC HYW007771500G
Encompass_Preferred
Acridone is an organic compound based on the acridine skeleton. It possesses antibacterial, antimalarial, antiviral, and antineoplastic activities.
  • Exhibits antibacterial, antimalarial, antiviral, and antineoplastic activities.
  • Inhibits the proliferation of MDA-MB-231 cancer cells (0.1-1 μM, 48 h) and blocks the mRNA and protein expression of ATP binding cassette subfamily G member 2 (ABCG2).
Catalog No. 50-004-47615