Learn More
Cayman Chemical Acriflavine Mixture, Purity: ≥95% (mixture of flavines), 5 mg
SDP
Catalog No. 502917543
Encompass_Preferred
A mixture of proflavine, acriflavine, side-methylated proflavine, and side-methylated acriflavine
Catalog No. 50-291-7543 Supplier Cayman Chemical Supplier No. 374075 MG
A mixture of proflavine, acriflavine, side-methylated proflavine, and side-methylated acriflavine