Learn More
Abcam Acriflavine hydrochloride, Fluorescent dye, 25G
Catalog No. 502704668
Encompass_Preferred
MW 468.97 g/mol. Fluorescent dye used as a biological stain and topical antiseptic. Mixture of 3,6-diaminoacridine (proflavin) and 3,6-diamino-10-methylacridinium (acriflavin). Has been used in applications for dual fluorescence analysis of cellular DNA and protein simultaneously.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-4668 Supplier Abcam Supplier No. AB14638325G
MW 468.97 g/mol. Fluorescent dye used as a biological stain and topical antiseptic. Mixture of 3,6-diaminoacridine (proflavin) and 3,6-diamino-10-methylacridinium (acriflavin). Has been used in applications for dual fluorescence analysis of cellular DNA and protein simultaneously.
The product is subject to the following: Abcam Restricted Use Statement