Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acriflavine Hydrochloride, Sigma-Aldrich

Catalog No. 01258955 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-258-955 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-258-955 Supplier MilliporeSigma Supelco Supplier No. A8251100G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Acriflavine is a fluorescent dye used for nuclear staining and microscopy.

  • Functions as a topical contrast agent for microscopy.
  • Used in confocal laser endomicroscopy of mucosal structure.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 8063-24-9

Specifications

Quantity 100 g
Product Type Dye
Label Type Other Labels or Dyes
Color Dark Orange
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.