Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acriflavine Hydrochloride, Spectrum™ Chemical
SDP

Catalog No. 18620155 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
5 kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18-620-155 5 kg
18-620-152 5 g
18-620-153 25 g
18-620-154 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18-620-155 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. AC1755KGBL
Get Up to $300 in Additional Chemicals
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 8063-24-9

Specifications

Packaging Drum
Quantity 5 kg
Molecular Weight (g/mol) 259.74
Grade Ungraded
Physical Form Powder or Waxy Solid
Chemical Name or Material Acriflavine Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.