Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acriflavine hydrochloride, Thermo Scientific Chemicals

Catalog No. AAJ6004814
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6004814 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ6004814 Supplier Thermo Scientific Chemicals Supplier No. J6004814
Add to Cart

missing translation for 'validMainframeMsg'

An intercalating dye

Acriflavine Hydrochloride acts as an intercalating dye and also reported to inhibit mitochondriogenesis. It is an antiseptic and has been used in applications for dual fluorescence analysis of cellular DNA and protein simultaneously.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Acriflavine Hydrochloride acts as an intercalating dye and also reported to inhibit mitochondriogenesis. It is an antiseptic and has been used in applications for dual fluorescence analysis of cellular DNA and protein simultaneously.

Solubility
Soluble in water, dimethyl sulfoxide and methanol.

Notes
A mixture of 3,6-diamino-10-methylacridinium chloride and 3,6-diaminoacridine
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 8063-24-9
Molecular Formula C27H27Cl3N6
Molecular Weight (g/mol) 541.91
MDL Number MFCD00069039
InChI Key MKLTXAHQKDVBLY-UHFFFAOYSA-N
Synonym 3,6-Diamino-10-methylacridinium chloride hydrochloride; Euflavine
IUPAC Name 3,6-diamino-10-methylacridin-10-ium acridine-3,6-diamine dihydrochloride chloride
SMILES Cl.Cl.[Cl-].NC1=CC2=NC3=CC(N)=CC=C3C=C2C=C1.C[N+]1=C2C=C(N)C=CC2=CC2=CC=C(N)C=C12

Specifications

Color Orange to Red
Quantity 25 g
UN Number UN3077
Beilstein 6265525
Merck Index 14,123
Solubility Information Soluble in water,dimethyl sulfoxide and methanol.
Physical Form Powder
Chemical Name or Material Acriflavine hydrochloride
Hazard Category H302-H318
Hazard Statement GHS H Statement
H318-H302
Causes serious eye damage.
Harmful if swallowed.
Precautionary Statement P264b-P270-P280-P301+P312-P305+P351+P338-P310-P330-P501c
DOTInformation Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.